Merck
  • Supramolecular Insights into Domino Effects of Ag@ZnO-Induced Oxidative Stress in Melanoma Cancer Cells.
Supramolecular Insights into Domino Effects of Ag@ZnO-Induced Oxidative Stress in Melanoma Cancer Cells.
ACS applied materials & interfaces (2019-11-16)
Behnaz Ghaemi, Arfa Moshiri, Inge K Herrmann, Mohammad Javad Hajipour, Peter Wick, Amir Amani, Sharmin Kharrazi
ABSTRACT
Recent studies suggest that cancer cell death accompanied by organelle dysfunction might be a promising approach for cancer therapy. The Golgi apparatus has a key role in cell function and may initiate signaling pathways to mitigate stress and, if irreparable, start apoptosis. It has been shown that Golgi disassembly and fragmentation under oxidative stress act as indicators for stress-mediated cell death pathways through cell cycle arrest in the G2/M phase. The present study shows that UV-induced reactive oxygen species (ROS) generation by Ag@ZnO nanoparticles (NPs) transform the Golgi structures from compressed perinuclear ribbons into detached vesicle-like structures distributed in the entire cytoplasm of melanoma cells. This study also demonstrates that Ag@ZnO NP-induced Golgi fragmentation cooccurs with G2 block of cell cycle progression, preventing cells from entering the mitosis phase. Additionally, the increased intracellular ROS production triggered by Ag@ZnO NPs upon UV exposure promoted autophagy. Taken together, Ag@ZnO NPs induce stress-related Golgi fragmentation and autophagy, finally leading to melanoma cell apoptosis. Intracellular oxidative stress generated by Ag@ZnO NPs upon UV irradiation may thus represent a targeted approach to induce cancer cell death through organelle destruction in melanoma cells, while fibroblast cells remained largely unaffected.
Sigma-Aldrich
2′,7′-Dichlorodihydrofluorescein diacetate, ≥97%
Sigma-Aldrich
Glutathione Reductase Assay Kit, Sufficient for 100 colorimetric tests