Merck
  • Efficient Semitransparent Organic Solar Cells with Tunable Color enabled by an Ultralow-Bandgap Nonfullerene Acceptor.
Efficient Semitransparent Organic Solar Cells with Tunable Color enabled by an Ultralow-Bandgap Nonfullerene Acceptor.
Advanced materials (Deerfield Beach, Fla.) (2017-10-05)
Yong Cui, Chenyi Yang, Huifeng Yao, Jie Zhu, Yuming Wang, Guoxiao Jia, Feng Gao, Jianhui Hou
ABSTRACT
Semitransparent organic solar cells (OSCs) show attractive potential in power-generating windows. However, the development of semitransparent OSCs is lagging behind opaque OSCs. Here, an ultralow-bandgap nonfullerene acceptor, "IEICO-4Cl", is designed and synthesized, whose absorption spectrum is mainly located in the near-infrared region. When IEICO-4Cl is blended with different polymer donors (J52, PBDB-T, and PTB7-Th), the colors of the blend films can be tuned from purple to blue to cyan, respectively. Traditional OSCs with a nontransparent Al electrode fabricated by J52:IEICO-4Cl, PBDB-T:IEICO-4Cl, and PTB7-Th:IEICO-4Cl yield power conversion efficiencies (PCE) of 9.65 ± 0.33%, 9.43 ± 0.13%, and 10.0 ± 0.2%, respectively. By using 15 nm Au as the electrode, semitransparent OSCs based on these three blends also show PCEs of 6.37%, 6.24%, and 6.97% with high average visible transmittance (AVT) of 35.1%, 35.7%, and 33.5%, respectively. Furthermore, via changing the thickness of Au in the OSCs, the relationship between the transmittance and efficiency is studied in detail, and an impressive PCE of 8.38% with an AVT of 25.7% is obtained, which is an outstanding value in the semitransparent OSCs.
Sigma-Aldrich
IEICO-4Cl, ≥99%
Pricing and availability is not currently available.