Merck
  • A fluorescent assay to quantitatively measure in vitro acyl CoA:diacylglycerol acyltransferase activity.
A fluorescent assay to quantitatively measure in vitro acyl CoA:diacylglycerol acyltransferase activity.
Journal of lipid research (2011-06-10)
Pamela J McFie, Scot J Stone
ABSTRACT
Triacylglycerols (TG) are the major storage form of energy in eukaryotic organisms and are synthesized primarily by acyl CoA:1,2-diacylglycerol acyltransferase (DGAT) enzymes. In vitro DGAT activity has previously been quantified by measuring the incorporation of either radiolabeled fatty acyl CoA or diacylglycerol (DG) into TG. We developed a modified acyltransferase assay using a fluorescent fatty acyl CoA substrate to accurately quantify in vitro DGAT activity. In the modified assay, radioactive fatty acyl CoA is replaced with fluorescent NBD-palmitoyl CoA, which is used as a substrate by DGAT with DG to produce NBD-TG. After extraction with organic solvents and separation by thin layer chromatography, NBD-TG formation can be detected and accurately quantified using a fluorescent imaging system. We demonstrate that this method can be adapted to detect other acyltransferase activities. Because NBD-palmitoyl CoA is commercially available at a much lower cost compared with radioactive acyl CoA substrates, it is a more economical alternative to radioactive tracers. In addition, the exposure of laboratory personnel to radioactivity is greatly reduced.
Sigma-Aldrich
1,2-Dioleoyl-sn-glycerol, ≥97%
Pricing and availability is not currently available.
Avanti
16-NBD-16:0 Coenzyme A, Avanti Research - A Croda Brand 810705P, powder
Pricing and availability is not currently available.