Merck
  • Integrative binding sites within intracellular termini of TRPV1 receptor.
Integrative binding sites within intracellular termini of TRPV1 receptor.
PloS one (2012-11-03)
Lenka Grycova, Blanka Holendova, Ladislav Bumba, Jan Bily, Michaela Jirku, Zdenek Lansky, Jan Teisinger
ABSTRACT
TRPV1 is a nonselective cation channel that integrates wide range of painful stimuli. It has been shown that its activity could be modulated by intracellular ligands PIP2 or calmodulin (CaM). The detailed localization and description of PIP2 interaction sites remain unclear. Here, we used synthesized peptides and purified fusion proteins of intracellular regions of TRPV1 expressed in E.coli in combination with fluorescence anisotropy and surface plasmon resonance measurements to characterize the PIP2 binding to TRPV1. We characterized one PIP2 binding site in TRPV1 N-terminal region, residues F189-V221, and two independent PIP2 binding sites in C-terminus: residues K688-K718 and L777-S820. Moreover we show that two regions, namely F189-V221 and L777-S820, overlap with previously localized CaM binding sites. For all the interactions the equilibrium dissociation constants were estimated. As the structural data regarding C-terminus of TRPV1 are lacking, restraint-based molecular modeling combined with ligand docking was performed providing us with structural insight to the TRPV1/PIP2 binding. Our experimental results are in excellent agreement with our in silico predictions.
Product Description
Avanti
16:0-06:0 NBD PC, Avanti Research - A Croda Brand 810130C
Avanti
16:0-06:0 NBD PC, Avanti Research - A Croda Brand 810130P, powder