Merck
  • The effects of zingerone against vancomycin-induced lung, liver, kidney and testis toxicity in rats: The behavior of some metabolic enzymes.
The effects of zingerone against vancomycin-induced lung, liver, kidney and testis toxicity in rats: The behavior of some metabolic enzymes.
Journal of biochemical and molecular toxicology (2019-08-28)
Cüneyt Çağlayan, Parham Taslimi, Yeliz Demir, Sefa Küçükler, Fatih Mehmet Kandemir, İlhami Gulçin
ABSTRACT
In this study, it was demonstrated the ameliorative effect of zingerone (ZO) (25 and 50 mg/kg body weight) against vancomycin (VCM) (200 mg/kg body weight) administered to rats on some metabolic enzymes' activities in the lung, liver, kidney, and testis tissues of rats. Forty-two rats were divided into six groups as follows: control, ZO-25, ZO-50, VCM, VCM + ZO-25, and VCM + ZO-50. α-Glycosidase, butyrylcholinesterase, aldose reductase, acetylcholinesterase, paraoxonase-1, and carbonic anhydrase enzyme activities were significantly (P < .05) decreased in VCM group when compared with the control group. ZO, supplied with VCM, significantly activated some of these enzyme in all tissues. The results of this study showed that ZO regulates abnormal increases and decreases in VCM-induced metabolic enzyme activities in all tissues.