Merck
  • Spread of ESBL-producing Escherichia coli in nursing home residents in Ireland and the Netherlands may reflect infrastructural differences.
Spread of ESBL-producing Escherichia coli in nursing home residents in Ireland and the Netherlands may reflect infrastructural differences.
The Journal of hospital infection (2019-05-12)
E M Terveer, M Fallon, M E M Kraakman, A Ormond, M Fitzpatrick, M A A Caljouw, A Martin, S M van Dorp, M C Wong, E J Kuijper, F Fitzpatrick
ABSTRACT
A prevalence study in two nursing homes (one each in the Netherlands and Ireland) found four (11%) Dutch and six (9%) Irish residents colonized with 11 extended-spectrum beta-lactamase-producing Escherichia coli, 10 of which contained CTX-M-15. Four Dutch isolates, from three residents of the same ward, belonged to E. coli O25:H4, sequence type (ST) 131 and were part of the same cluster type by whole-genome sequencing. Four Irish residents on three different wards were colonized with an identical E. coli O89:H9, ST131, complex type 1478. Cross-transmission between three Irish wards may reflect differences in nursing home infrastructure, specifically communal areas and multi-bedded resident rooms.
Sigma-Aldrich
5-Norbornene-2-endo,3-endo-dimethanol, 98%
Pricing and availability is not currently available.