Merck
Bio-inspired hydrophobicity promotes CO2 reduction on a Cu surface.
Nature materials (2019-08-07)
David Wakerley, Sarah Lamaison, François Ozanam, Nicolas Menguy, Dimitri Mercier, Philippe Marcus, Marc Fontecave, Victor Mougel
ABSTRACT
The aqueous electrocatalytic reduction of CO2 into alcohol and hydrocarbon fuels presents a sustainable route towards energy-rich chemical feedstocks. Cu is the only material able to catalyse the substantial formation of multicarbon products (C2/C3), but competing proton reduction to hydrogen is an ever-present drain on selectivity. Here, a superhydrophobic surface was generated by 1-octadecanethiol treatment of hierarchically structured Cu dendrites, inspired by the structure of gas-trapping cuticles on subaquatic spiders. The hydrophobic electrode attained a 56% Faradaic efficiency for ethylene and 17% for ethanol production at neutral pH, compared to 9% and 4% on a hydrophilic, wettable equivalent. These observations are assigned to trapped gases at the hydrophobic Cu surface, which increase the concentration of CO2 at the electrode-solution interface and consequently increase CO2 reduction selectivity. Hydrophobicity is thus proposed as a governing factor in CO2 reduction selectivity and can help explain trends seen on previously reported electrocatalysts.
Sigma-Aldrich
1-Octadecanethiol, 98%