Merck
  • Hydrophobic scaffolds of pH-sensitive cationic lipids contribute to miscibility with phospholipids and improve the efficiency of delivering short interfering RNA by small-sized lipid nanoparticles.
Hydrophobic scaffolds of pH-sensitive cationic lipids contribute to miscibility with phospholipids and improve the efficiency of delivering short interfering RNA by small-sized lipid nanoparticles.
Acta biomaterialia (2019-11-17)
Yusuke Sato, Nana Okabe, Yusuke Note, Kazuki Hashiba, Masatoshi Maeki, Manabu Tokeshi, Hideyoshi Harashima
ABSTRACT
Despite the fact that small-sized lipid nanoparticles (LNPs) are important for improved tissue penetration and efficient drug delivery, their poor stability and intracellular trafficking significantly hinders their use as potent small-sized LNPs. It has been reported that both the diffusion of lipid components from LNPs and the adsorption of proteins on the surface of LNPs are responsible for their decreased potency. To overcome this issue, we focused on the chemical structure of hydrophobic scaffolds of pH-sensitive cationic lipids with various lengths and shapes. LNPs composed of a pH-sensitive cationic lipid with long, linear scaffolds induced gene silencing in a dose-dependent manner, while LNPs with a classical scaffold length (C18) failed. Replacing the helper lipid from cholesterol to egg sphingomyelin (ESM) resulted in the formation of smaller LNPs with a diameter of ~22 nm and enhanced gene silencing activity. Most of the ESMs were located in the outer layer and functioned to stabilize the LNPs. Long, linear scaffolds contributed to immiscibility with phosphocholine-containing lipids including ESM. This contribution was dependent on the scaffold length of pH-sensitive cationic lipids. Although phosphocholine-containing lipids usually inhibit membrane fusion-mediated endosomal escape, long, linear scaffolds contributed to avoiding the inhibitory effect and to enhance the potency of the LNPs. These findings provide useful information needed for the rational design of pH-sensitive cationic lipid structures and the selection of appropriate helper lipids and will facilitate the development of highly potent small-sized LNPs. STATEMENT OF SIGNIFICANCE: Despite the fact that small-sized lipid nanoparticles (LNPs) are important for improved tissue penetration and efficient drug delivery, the size reduction-associated decrease in the stability and intracellular trafficking significantly hinders the development of potent small-sized LNPs. Our limited understanding of the mechanism underlying the reduced potency has also hindered the development of more potent small-sized LNPs. The findings of the present study indicate that long and linear hydrophobic scaffolds of pH-sensitive cationic lipids could overcome the loss of efficiency for nucleic acid delivery. In addition, the long hydrophobic scaffolds led to immiscibility with neutral phospholipids, resulting in efficient endosomal escape. These findings provide useful information needed for the rational design of pH-sensitive cationic lipid structures and will facilitate the development of highly potent small-sized LNPs.
Sigma-Aldrich
Triton X-100, laboratory grade
Pricing and availability is not currently available.
Avanti
Brain SM, Avanti Research - A Croda Brand
Pricing and availability is not currently available.
Avanti
Brain SM, Avanti Research - A Croda Brand
Pricing and availability is not currently available.