Merck
  • Structure-Solubility Relationship of 1,4-Dioxane Complexes of Di(hydrocarbyl)magnesium.
Structure-Solubility Relationship of 1,4-Dioxane Complexes of Di(hydrocarbyl)magnesium.
Chemistry (Weinheim an der Bergstrasse, Germany) (2019-07-23)
Reinald Fischer, Helmar Görls, Philippe R Meisinger, Regina Suxdorf, Matthias Westerhausen
ABSTRACT
Systematic variation of the 1,4-dioxane (dx) concentration during the precipitation of sparingly soluble [MgBr2 (dx)2 ] from ethereal Grignard solutions of RMgBr has allowed the structural investigation of crystallized [R2 Mg(dx)n ] (n=1, 1.5, 2, and 3), which form during this dioxane method, depending on the bulkiness of R. The numbering of the complexes explored in this study is based on the number n of dioxane molecules per magnesium atom, followed by the substituent R; an apostrophe denotes coordination polymers. The following derivatives were studied by X-ray crystal-structure determination and NMR spectroscopy: n=1: [Me2 Mg(μ-dx)]∞ (1'-Me) and [nPr2 Mg(μ-dx)]∞ (1'-nPr); n=1.5: [{iPr2 Mg(dx)}2 (μ-dx)] (1.5-iPr), [{oTol2 Mg(dx)}2 (μ-dx)] (1.5-oTol), and [(Me3 Si-C≡C)2 Mg(dx)1.5 ]∞ (1.5'-C2 SiMe3 ); n=2: [tBu2 Mg(dx)2 ] (2-tBu) and [oTol2 Mg(dx)2 ] (2-oTol); n=3: [Ph2 Mg(dx)3 ] (3-Ph). In the structure types 1', 1.5, and 2, the magnesium atom exhibits the coordination number 4, whereas pentacoordinate metal atoms are observed in types 3 and 1.5'. The structure type 2' is realized for [(Ph-C≡C)2 Mg(dx)2 ]∞ (2'-C2 Ph), [MgCl2 (dx)2 ]∞ (2'-Cl), and [MgBr2 (dx)2 ]∞ (2'-Br) with hexacoordinate metal atoms. The solubility of the dioxane adducts in common organic solvents strongly depends on the degree of aggregation with the solubility decreasing from molecular to strand to layer structures.
Sigma-Aldrich
2-Bromotoluene, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethynyltrimethylsilane, 98%
Pricing and availability is not currently available.