- Double-Bond Isomerization: Highly Reactive Nickel Catalyst Applied in the Synthesis of the Pheromone (9Z,12Z)-Tetradeca-9,12-dienyl Acetate.
Double-Bond Isomerization: Highly Reactive Nickel Catalyst Applied in the Synthesis of the Pheromone (9Z,12Z)-Tetradeca-9,12-dienyl Acetate.
Organic letters (2015-05-30)
Felicia Weber, Anastasia Schmidt, Philipp Röse, Michel Fischer, Olaf Burghaus, Gerhard Hilt
PMID26023712
ABSTRACT
A highly reactive nickel catalyst comprising NiCl2(dppp) or NiCl2(dppe) with zinc powder, ZnI2 and Ph2PH, was applied in the isomerization of terminal alkenes to Z-2-alkenes. The double-bond geometry of the 2-alkene can be controlled via the reaction temperature to yield the 2-Z-alkenes in excellent yields and high Z-selectivities. The formation of other constitutional isomers, such as 3-alkenes, is suppressed on the basis of the proposed mechanism via a 1,2-hydride shift from the metal to the Ph2P ligand. The nickel-catalyzed isomerization reaction was then applied in the synthesis of (9Z,12Z)-tetradeca-9,12-dienyl acetate, a pheromone with a 2Z,5Z-diene subunit.