Merck
  • Norepinephrine-Induced Stimulation of Kir4.1/Kir5.1 Is Required for the Activation of NaCl Transporter in Distal Convoluted Tubule.
Norepinephrine-Induced Stimulation of Kir4.1/Kir5.1 Is Required for the Activation of NaCl Transporter in Distal Convoluted Tubule.
Hypertension (Dallas, Tex. : 1979) (2018-12-21)
Xin-Peng Duan, Li Gu, Yu Xiao, Zhong-Xiuzi Gao, Peng Wu, Yun-Hong Zhang, Xin-Xin Meng, Jun-Lin Wang, Dan-Dan Zhang, Dao-Hong Lin, Wen-Hui Wang, Ruimin Gu
ABSTRACT
The stimulation of β-adrenergic receptor increases thiazide-sensitive NaCl cotransporter (NCC), an effect contributing to salt-sensitive hypertension by sympathetic stimulation. We now test whether the stimulation of β-adrenergic receptor-induced activation of NCC is achieved through activating basolateral Kir4.1 in the distal convoluted tubule (DCT). Application of norepinephrine increased the basolateral 40 pS K+ channel (Kir4.1/Kir5.1 heterotetramer) in the DCT. The stimulatory effect of norepinephrine on the K+ channel was mimicked by cAMP analogue but abolished by inhibiting PKA (protein kinase A). Also, the effect of norepinephrine on the K+ channel in the DCT was recapitulated by isoproterenol but not by α-adrenergic agonist and blocked by propranolol, suggesting that norepinephrine effect on the K+ channel was mediated by β-adrenergic receptor. The whole-cell recording shows that norepinephrine and isoproterenol increased DCT K+ currents and shifted the K+ current ( IK) reversal potential to negative range (hyperpolarization). Continuous norepinephrine perfusion (7 days) increased DCT K+ currents, hyperpolarized IK reversal potential, and increased the expression of total NCC/phosphorylated NCC, but it had no significant effect on the expression of NKCC2 (type 2 Na-Cl-K cotransporter) and ENaC-α (epithelial Na channel-α subunit). Renal clearance study demonstrated that norepinephrine perfusion augmented thiazide-induced urinary Na+ excretion only in wild-type but not in kidney-specific Kir4.1 knockout mice, suggesting that Kir4.1 is required for mediating the effect of norepinephrine on NCC. However, norepinephrine perfusion did not affect urinary K+ excretion. We conclude that the stimulation of β-adrenergic receptor activates the basolateral Kir4.1 in the DCT and that the activation of Kir4.1 is required for norepinephrine-induced stimulation of NCC.
Sigma-Aldrich
Anti-Thiazide-Sensitive NaCl Cotransporter Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-SCNN1A antibody produced in rabbit, 1 mg/mL, affinity isolated antibody