Merck
  • Abrogation of cathepsin C by Helicobacter pylori impairs neutrophil activation to promote gastric infection.
Abrogation of cathepsin C by Helicobacter pylori impairs neutrophil activation to promote gastric infection.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2019-01-01)
Yu Gang Liu, Yong Sheng Teng, Ping Cheng, Hui Kong, Yi Pin Lv, Fang Yuan Mao, Xiao Long Wu, Chuan Jie Hao, Weisan Chen, Shi Ming Yang, Jin Yu Zhang, Liu Sheng Peng, Ting Ting Wang, Bin Han, Qiang Ma, Quan Ming Zou, Yuan Zhuang
ABSTRACT
Cathepsin C (CtsC) functions as a central coordinator for activation of many serine proteases in immune cells. However, CtsC expression in gastric epithelial cells and its role in Helicobacter pylori infection remain unclear. Real-time PCR, Western blot, and immunohistochemistry analyses identified that CtsC was decreased in gastric mucosa of H. pylori-infected patients and mice. Isolated gastric epithelial cells and cell lines were stimulated with H. pylori and/or TGF-β1 showed that down-regulation of CtsC in gastric epithelial cells largely depended on H. pylori cagA via Src/ERK and Janus kinase (JAK)/signal transducer and activator of transcription 3 (STAT3) pathways, and the effect could be synergistically augmented by TGF-β1 in an autocrine manner. In human gastric mucosa, CtsC expression was negatively correlated with bacteria colonization; accordingly, provision of exogenous active CtsC overwhelmed H. pylori persistence in gastric mucosa of mice. In the presence of active CtsC, isolated human neutrophils activated via NF-κB pathway with augmented bactericidal capacity in vitro. We also found that neutrophils activated and cleared bacteria in active CtsC-injected mice and that there was no bactericidal capacity in mice that were simultaneously neutrophil-depleted by Ly6G antibody. Our findings identified a mechanism that H. pylori abrogate CtsC to impair neutrophil activation and to ensure persistence in gastric mucosa. Efforts to enable and boost this neutrophil activation pathway by active CtsC may therefore become valuable strategies in treating H. pylori infection.-Liu, Y. G., Teng, Y. S., Cheng, P., Kong, H., Lv, Y. P., Mao, F. Y., Wu, X. L., Hao, C. J., Chen, W., Yang, S. M., Zhang, J. Y., Peng, L. S., Wang, T. T., Han, B., Ma, Q., Zou, Q. M., Zhuang, Y. Abrogation of cathepsin C by Helicobacter pylori impairs neutrophil activation to promote gastric infection.
Sigma-Aldrich
JNK Inhibitor II, JNK Inhibitor II. SP600125, CAS 129-56-6, is a potent, cell-permeable, selective, and ATP competitive inhibitor of c-Jun N-terminal kinase (JNK; IC₅₀ = 40 nM for JNK-1 & JNK-2 & 90 nM for JNK-3).
Sigma-Aldrich
PP2, PP2, CAS 172889-27-9, is a potent, reversible, ATP-competitive, inhibitor of the Src family of protein tyrosine kinases (IC₅₀ = 4, 5, 5, &100 nM for p56lck, p59fynT, Hck, & Src, respectively).
Sigma-Aldrich
GSK-3β Inhibitor VI, The GSK-3β Inhibitor VI, also referenced under CAS 62673-69-2, controls the biological activity of GSK-3β. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
U0126, U0126 is a elective inhibitor of MEK-1 and MEK-2; weak inhibitor of PKC, Raf, ERK, JNK, MEKK, MKK-3, MKK-4/SEK, MKK-6, Abl, Cdk2 and Cdk4.