Merck
  • Acidic Stress Triggers Sodium-Coupled Bicarbonate Transport and Promotes Survival in A375 Human Melanoma Cells.
Acidic Stress Triggers Sodium-Coupled Bicarbonate Transport and Promotes Survival in A375 Human Melanoma Cells.
Scientific reports (2019-05-03)
Oscar C Y Yang, Shih-Hurng Loh
ABSTRACT
Melanoma cells preserve intracellular pH (pHi) within a viable range despite an acidic ambient pH that typically falls below pH 7.0. The molecular mechanisms underlying this form of acidic preservation in melanoma remain poorly understood. Previous studies had demonstrated that proton transporters including the monocarboxylate transporter (MCT), the sodium hydrogen exchanger (NHE), and V-Type ATPase mediate acid extrusion to counter intracellular acidification in melanoma cells. In this report, the expression and function of the Sodium-Coupled Bicarbonate Transporter (NCBT) family of base loaders were further characterized in melanoma cell lines. NCBT family members were found to be expressed in three different melanoma cell lines - A375, MeWo, and HS695T - and included the electrogenic sodium-bicarbonate cotransporter isoforms 1 and 2 (NBCe1 and NBCe2), the electroneutral sodium-bicarbonate cotransporter (NBCn1), and the sodium-dependent chloride-bicarbonate exchanger (NDCBE). These transporters facilitated 4,4'-diisothiocyanatostilbene-2,2'-disulfonic acid (DIDS)-dependent pHi recovery in melanoma cells, in response to intracellular acidification induced by ammonium chloride prepulse. Furthermore, the expression of NCBTs were upregulated via chronic exposure to extracellular acidification. Given the current research interest in the NCBTs as a molecular driver of tumourigenesis, characterising NCBT in melanoma provides impetus for developing novel therapeutic targets for melanoma treatment.