Merck
  • 23-Hydroxyursolic Acid Isolated from the Stem Bark of Cussonia bancoensis Induces Apoptosis through Fas/Caspase-8-Dependent Pathway in HL-60 Human Promyelocytic Leukemia Cells.
23-Hydroxyursolic Acid Isolated from the Stem Bark of Cussonia bancoensis Induces Apoptosis through Fas/Caspase-8-Dependent Pathway in HL-60 Human Promyelocytic Leukemia Cells.
Molecules (Basel, Switzerland) (2018-12-16)
Jong-Heon Won, Kyung-Sook Chung, Eun-Young Park, Jeong-Hun Lee, Jung-Hye Choi, Leon Azefack Tapondjou, Hee-Juhn Park, Masaaki Nomura, Ahmed H E Hassan, Kyung-Tae Lee
ABSTRACT
The natural product 23-hydroxyursolic acid (23-HUA) is a derivative of ursolic acid, which is known to induce cancer cell apoptosis. However, apoptotic effects and mechanisms of 23-HUA have not been well characterized yet. Herein, we investigated the molecular mechanisms of 23-HUA-induced apoptosis in HL-60 human promyelocytic leukemia cells. 23-HUA-treated HL-60 cells showed apoptotic features including internucleosomal DNA condensation and fragmentation as well as externalization of phosphatidylserine residues. 23-HUA induced a series of mitochondrial events including disruption of mitochondrial membrane potential (ΔΨm), cytochrome c and Smac/DIABLO release and loss of balance between pro-apoptotic and anti-apoptotic Bcl-2 proteins in HL-60 cells. In addition, 23-HUA activated caspase-8, caspase-9 and caspase-3. Pretreatment with a broad caspase inhibitor (z-VAD-fmk), a caspase-3 inhibitor (z-DEVD-fmk), and a caspase-8 inhibitor (z-IETD-fmk) significantly attenuated 23-HUA-induced DNA fragmentation. After 23-HUA-induced apoptosis, proteins expression levels of FasL, Fas and FADD constituting the death-inducing signaling complex (DISC) were upregulated in HL-60 cells. Moreover, transfection with Fas or FADD siRNA significantly blocked 23-HUA-induced DNA fragmentation and caspases activation. Taken together, these findings indicate that 23-HUA induces apoptosis in HL-60 human promyelocytic leukemia cells through formation of DISC and caspase-8 activation leading to loss of ΔΨm and caspase-3 activation.
Sigma-Aldrich
Z-VAD-FMK, solid