Merck
  • MicroRNA-98 attenuates cardiac ischemia-reperfusion injury through inhibiting DAPK1 expression.
MicroRNA-98 attenuates cardiac ischemia-reperfusion injury through inhibiting DAPK1 expression.
IUBMB life (2018-11-13)
Chang-Lin Zhai, Guan-Min Tang, Gang Qian, Hui-Lin Hu, Shi-Jun Wang, Dong Yin, Song Zhang
ABSTRACT
Cardiovascular ischemic disease is a large class of diseases that are harmful to human health. The significant role of microRNAs (miRNAs) in terms of controlling cardiac injury has been reported in latest studies. MiR-98 is very important in regulating the apoptosis, the differentiation, the growth as well as the metastasis of cells. Nevertheless, the effect of miR-98 in the cardiac ischemia reperfusion (I/R) injury has rarely been investigated. In the current research, we found that the miR-98 expression was down-regulated in the cardiomyocytes subjected to hypoxia/reoxygenation (H/R) and in the myocardium of the I/R rats. In addition, over-expression of miR-98 could significantly reduce the myocardial oxidative stress and ischemic injury as well as cell apoptosis. In agreement, similar findings were demonstrated in H9c2 cells subjected to H/R injury. Bioinformatic analysis using MiRanda and TargetScan and luciferase activity assay confirmed death-associated protein kinase 1 (DAPK1) as a direct target of miR-98. These findings suggest that miR-98 may be exploited as a novel molecular marker or therapeutic target for myocardial I/R injury. © 2018 IUBMB Life, 71(1):166-176, 2019.