Merck
  • A selective ER-phagy exerts procollagen quality control via a Calnexin-FAM134B complex.
A selective ER-phagy exerts procollagen quality control via a Calnexin-FAM134B complex.
The EMBO journal (2018-12-19)
Alison Forrester, Chiara De Leonibus, Paolo Grumati, Elisa Fasana, Marilina Piemontese, Leopoldo Staiano, Ilaria Fregno, Andrea Raimondi, Alessandro Marazza, Gemma Bruno, Maria Iavazzo, Daniela Intartaglia, Marta Seczynska, Eelco van Anken, Ivan Conte, Maria Antonietta De Matteis, Ivan Dikic, Maurizio Molinari, Carmine Settembre
ABSTRACT
Autophagy is a cytosolic quality control process that recognizes substrates through receptor-mediated mechanisms. Procollagens, the most abundant gene products in Metazoa, are synthesized in the endoplasmic reticulum (ER), and a fraction that fails to attain the native structure is cleared by autophagy. However, how autophagy selectively recognizes misfolded procollagens in the ER lumen is still unknown. We performed siRNA interference, CRISPR-Cas9 or knockout-mediated gene deletion of candidate autophagy and ER proteins in collagen producing cells. We found that the ER-resident lectin chaperone Calnexin (CANX) and the ER-phagy receptor FAM134B are required for autophagy-mediated quality control of endogenous procollagens. Mechanistically, CANX acts as co-receptor that recognizes ER luminal misfolded procollagens and interacts with the ER-phagy receptor FAM134B. In turn, FAM134B binds the autophagosome membrane-associated protein LC3 and delivers a portion of ER containing both CANX and procollagen to the lysosome for degradation. Thus, a crosstalk between the ER quality control machinery and the autophagy pathway selectively disposes of proteasome-resistant misfolded clients from the ER.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-HA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-FAM134B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody