Merck
  • Immature excitatory neurons develop during adolescence in the human amygdala.
Immature excitatory neurons develop during adolescence in the human amygdala.
Nature communications (2019-06-23)
Shawn F Sorrells, Mercedes F Paredes, Dmitry Velmeshev, Vicente Herranz-Pérez, Kadellyn Sandoval, Simone Mayer, Edward F Chang, Ricardo Insausti, Arnold R Kriegstein, John L Rubenstein, Jose Manuel Garcia-Verdugo, Eric J Huang, Arturo Alvarez-Buylla
ABSTRACT
The human amygdala grows during childhood, and its abnormal development is linked to mood disorders. The primate amygdala contains a large population of immature neurons in the paralaminar nuclei (PL), suggesting protracted development and possibly neurogenesis. Here we studied human PL development from embryonic stages to adulthood. The PL develops next to the caudal ganglionic eminence, which generates inhibitory interneurons, yet most PL neurons express excitatory markers. In children, most PL cells are immature (DCX+PSA-NCAM+), and during adolescence many transition into mature (TBR1+VGLUT2+) neurons. Immature PL neurons persist into old age, yet local progenitor proliferation sharply decreases in infants. Using single nuclei RNA sequencing, we identify the transcriptional profile of immature excitatory neurons in the human amygdala between 4-15 years. We conclude that the human PL contains excitatory neurons that remain immature for decades, a possible substrate for persistent plasticity at the interface of the hippocampus and amygdala.
Sigma-Aldrich
Anti-Nitric Oxide Synthase I Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Doublecortin Antibody, serum, from guinea pig
Sigma-Aldrich
Anti-Polysialic Acid-NCAM Antibody, clone 2-2B, ascites fluid, clone 2-2B, Chemicon®
Sigma-Aldrich
Anti-NeuN Antibody, from chicken, purified by affinity chromatography
Sigma-Aldrich
Anti-Tbr1 Antibody, from chicken, purified by affinity chromatography
Sigma-Aldrich
Anti-Sox11 Antibody, pain, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Monoclonal Anti-Vimentin antibody produced in mouse, clone VIM-13.2, ascites fluid
Sigma-Aldrich
Anti-SCGN antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Brain lipid binding protein Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-Prox 1 Antibody, serum, Chemicon®