Merck
  • JARID2 Functions as a Tumor Suppressor in Myeloid Neoplasms by Repressing Self-Renewal in Hematopoietic Progenitor Cells.
JARID2 Functions as a Tumor Suppressor in Myeloid Neoplasms by Repressing Self-Renewal in Hematopoietic Progenitor Cells.
Cancer cell (2018-11-14)
Hamza Celik, Won Kyun Koh, Ashley C Kramer, Elizabeth L Ostrander, Cates Mallaney, Daniel A C Fisher, Jingyu Xiang, William C Wilson, Andrew Martens, Alok Kothari, Gregory Fishberger, Eric Tycksen, Darja Karpova, Eric J Duncavage, Youngsook Lee, Stephen T Oh, Grant A Challen
ABSTRACT
How specific genetic lesions contribute to transformation of non-malignant myeloproliferative neoplasms (MPNs) and myelodysplastic syndromes (MDSs) to secondary acute myeloid leukemia (sAML) are poorly understood. JARID2 is lost by chromosomal deletions in a proportion of MPN/MDS cases that progress to sAML. In this study, genetic mouse models and patient-derived xenografts demonstrated that JARID2 acts as a tumor suppressor in chronic myeloid disorders. Genetic deletion of Jarid2 either reduced overall survival of animals with MPNs or drove transformation to sAML, depending on the timing and context of co-operating mutations. Mechanistically, JARID2 recruits PRC2 to epigenetically repress self-renewal pathways in hematopoietic progenitor cells. These studies establish JARID2 as a bona fide hematopoietic tumor suppressor and highlight potential therapeutic targets.
Sigma-Aldrich
Normal Rabbit IgG, Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Pricing and availability is not currently available.
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Pricing and availability is not currently available.