- Isolation and structure determination of a new cytotoxic peptide, curacozole, from Streptomyces curacoi based on genome mining.
Isolation and structure determination of a new cytotoxic peptide, curacozole, from Streptomyces curacoi based on genome mining.
The Journal of antibiotics (2018-10-13)
Issara Kaweewan, Hisayuki Komaki, Hikaru Hemmi, Kanata Hoshino, Takeshi Hosaka, Gouchi Isokawa, Takanori Oyoshi, Shinya Kodani
PMID30310179
ABSTRACT
Using genome mining, a new cytotoxic peptide named curacozole was isolated from Streptomyces curacoi. Through ESI-MS and NMR analyses, curacozole was determined to be a macrocyclic peptide containing two isoleucine, two thiazole and three oxazole moieties. Curacozole exhibited potent cytotoxic activity against HCT116 and HOS cancer cells. The proposed biosynthetic gene cluster of curacozole was identified and compared with that of the related compound YM-216391.