Merck
  • Chitosan oligosaccharide-based dual pH responsive nano-micelles for targeted delivery of hydrophobic drugs.
Chitosan oligosaccharide-based dual pH responsive nano-micelles for targeted delivery of hydrophobic drugs.
Carbohydrate polymers (2019-08-21)
Yulong Wang, Avik Khan, Yanxin Liu, Jing Feng, Lei Dai, Guanhua Wang, Nur Alam, Li Tong, Yonghao Ni
ABSTRACT
In this study, chitosan oligosaccharide (COS)-vanillin imine (COS-Vani Imine)-based dual pH responsive nano-micelles (DPRNs) were synthesized. The resultant DPRNs were used for encapsulating genistein and its ultimate release upon pH change. The overall concept of DPRNs for the targeted delivery of hydrophobic anticancer drugs was successfully demonstrated. The DPRNs were spherical in shape, nanoscale in dimension (71.2-163.4 nm), with dual pH response. The encapsulation/loading of genistein into DPRNs was achieved and the resultant genistein-loaded DPRNs were stable under the physiological pH (˜7.4); under the cancer cell extracellular pH (˜6.8), the amino groups in COS is protonated, thus becoming positively charged, facilitating their adsorption onto negatively charged cancer cells. Under the cancer cell intracellular pH (˜5.0), the genistein-loaded DPRNs were destroyed as a result of the cleavage of pH sensitive benzoic imine, thereby releasing the loaded genistein.