Merck
  • PARP1 Trapping by PARP Inhibitors Drives Cytotoxicity in Both Cancer Cells and Healthy Bone Marrow.
PARP1 Trapping by PARP Inhibitors Drives Cytotoxicity in Both Cancer Cells and Healthy Bone Marrow.
Molecular cancer research : MCR (2018-11-16)
Todd A Hopkins, William B Ainsworth, Paul A Ellis, Cherrie K Donawho, Enrico L DiGiammarino, Sanjay C Panchal, Vivek C Abraham, Mikkel A Algire, Yan Shi, Amanda M Olson, Eric F Johnson, Julie L Wilsbacher, David Maag
ABSTRACT
PARP inhibitors have recently been approved as monotherapies for the treatment of recurrent ovarian cancer and metastatic BRCA-associated breast cancer, and ongoing studies are exploring additional indications and combinations with other agents. PARP inhibitors trap PARP onto damaged chromatin when combined with temozolomide and methyl methanesulfonate, but the clinical relevance of these findings remains unknown. PARP trapping has thus far been undetectable in cancer cells treated with PARP inhibitors alone. Here, we evaluate the contribution of PARP trapping to the tolerability and efficacy of PARP inhibitors in the monotherapy setting. We developed a novel implementation of the proximity ligation assay to detect chromatin-trapped PARP1 at single-cell resolution with higher sensitivity and throughput than previously reported methods. We further demonstrate that the PARP inhibitor-induced trapping appears to drive single-agent cytotoxicity in healthy human bone marrow, indicating that the toxicity of trapped PARP complexes is not restricted to cancer cells with homologous recombination deficiency. Finally, we show that PARP inhibitors with dramatically different trapping potencies exhibit comparable tumor growth inhibition at MTDs in a xenograft model of BRCA1-mutant triple-negative breast cancer. These results are consistent with emerging clinical data and suggest that the inverse relationship between trapping potency and tolerability may limit the potential therapeutic advantage of potent trapping activity. IMPLICATIONS: PARP trapping contributes to single-agent cytotoxicity of PARP inhibitors in both cancer cells and healthy bone marrow, and the therapeutic advantage of potent trapping activity appears to be limited.