- Efforts toward distorted spiropentanes.
Efforts toward distorted spiropentanes.
The Journal of organic chemistry (2010-10-14)
Kuan-Jen Su, Jean-Luc Mieusset, Vladimir B Arion, Wolfgang Knoll, Lothar Brecker, Udo H Brinker
PMID20939558
ABSTRACT
Tetravinylbenzene 4 was prepared in nearly quantitative yield from commercially available tetrabromobenzene; the improved, one-step procedure now employs Suzuki-Miyaura cross-coupling conditions. Intermolecular cyclopropanation of 4 with dibromocarbene gave a series of gem-dibromide adducts. Intramolecular cyclopropanation of monoadduct 5, putatively by its methyllithium-generated cyclopropylidene(oid), produced compound 11, which features a highly distorted spiropentane having a C-C-C bond angle of 163.5°. The stability of the reported spiropentanes was investigated using DFT calculations.