Merck
  • Pitfalls of polymyxin antimicrobial susceptibility testing of Pseudomonas aeruginosa isolated from cystic fibrosis patients.
Pitfalls of polymyxin antimicrobial susceptibility testing of Pseudomonas aeruginosa isolated from cystic fibrosis patients.
The Journal of antimicrobial chemotherapy (2004-10-29)
Michael Hogardt, Sabine Schmoldt, Monika Götzfried, Kristin Adler, Jürgen Heesemann
ABSTRACT
With their potent activity against Gram-negative bacteria, the polymyxins are important alternative antibiotics for cystic fibrosis (CF) patients. A retrospective evaluation of polymyxin activity against 6001 Pseudomonas aeruginosa, 150 Achromobacter xylosoxidans and 506 Stenotrophomonas maltophilia CF isolates was initiated. In addition, we looked at how polymyxin susceptibility testing was affected by the testing method (agar dilution versus microdilution), the agent (polymyxin E versus polymyxin B), incubation time (24 h versus 48 h) and by different interpretative criteria (German DIN, French FSM, British BSAC). Polymyxin B exhibited reasonable activity against P. aeruginosa (MIC(90)< or =2 mg/L), whereas it was less active against A. xylosoxidans (MIC(90)< or =16 mg/L) and S. maltophilia (MIC(90)< or =16 mg/L). During 2000-2002, polymyxin B resistance in P. aeruginosa, S. maltophilia and A. xylosoxidans was found to be 6.7%, 17.0% and 29.9% (corresponding to 12.4%, 20.7% and 35.4% of infected patients), respectively. When the agar dilution method was used, polymyxin E exhibited higher MICs than polymyxin B. The microdilution method produced lower polymyxin MICs than the agar dilution method. Therefore, the microdilution MICs after prolonged incubation (48 h) and the agar dilution MICs of polymyxin B correlated best (AUC of 0.93, r(2) of 0.44 and s of 0.83). Polymyxin resistance among common CF pathogens is not rare, thus underlining the necessity of accurate susceptibility testing. When compared with the agar dilution method, it was found that the microdilution method is a valid, rapid and cost effective alternative for the determination of polymyxin activity. The performance of the microdilution method was most reliable after prolonged incubation (48 h) at a susceptibility breakpoint of < or =4 mg/L according to the BSAC guidelines (specificity 91%, sensitivity 89%, 1.5% very major errors).
Sigma-Aldrich
Polymyxin B solution, 20 mg/mL in H2O
Pricing and availability is not currently available.