Merck
  • Characterization of GSK'963: a structurally distinct, potent and selective inhibitor of RIP1 kinase.
Characterization of GSK'963: a structurally distinct, potent and selective inhibitor of RIP1 kinase.
Cell death discovery (2015-01-01)
S B Berger, P Harris, R Nagilla, V Kasparcova, S Hoffman, B Swift, L Dare, M Schaeffer, C Capriotti, M Ouellette, B W King, D Wisnoski, J Cox, M Reilly, R W Marquis, J Bertin, P J Gough
ABSTRACT
Necroptosis and signaling regulated by RIP1 kinase activity is emerging as a key driver of inflammation in a variety of disease settings. A significant amount has been learned about how RIP1 regulates necrotic cell death through the use of the RIP1 kinase inhibitor Necrostatin-1 (Nec-1). Nec-1 has been a transformational tool for exploring the function of RIP1 kinase activity; however, its utility is somewhat limited by moderate potency, off-target activity against indoleamine-2,3-dioxygenase (IDO), and poor pharmacokinetic properties. These limitations of Nec-1 have driven an effort to identify next-generation tools to study RIP1 function, and have led to the identification of 7-Cl-O-Nec-1 (Nec-1s), which has improved pharmacokinetic properties and lacks IDO inhibitory activity. Here we describe the characterization of GSK'963, a chiral small-molecule inhibitor of RIP1 kinase that is chemically distinct from both Nec-1 and Nec-1s. GSK'963 is significantly more potent than Nec-1 in both biochemical and cellular assays, inhibiting RIP1-dependent cell death with an IC50 of between 1 and 4 nM in human and murine cells. GSK'963 is >10 000-fold selective for RIP1 over 339 other kinases, lacks measurable activity against IDO and has an inactive enantiomer, GSK'962, which can be used to confirm on-target effects. The increased in vitro potency of GSK'963 also translates in vivo, where GSK'963 provides much greater protection from hypothermia at matched doses to Nec-1, in a model of TNF-induced sterile shock. Together, we believe GSK'963 represents a next-generation tool for examining the function of RIP1 in vitro and in vivo, and should help to clarify our current understanding of the role of RIP1 in contributing to disease pathogenesis.
Sigma-Aldrich
GSK′962, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
GSK′963, ≥98% (HPLC)
Pricing and availability is not currently available.