Merck
  • Pharmacologic inhibition of the anaphase-promoting complex induces a spindle checkpoint-dependent mitotic arrest in the absence of spindle damage.
Pharmacologic inhibition of the anaphase-promoting complex induces a spindle checkpoint-dependent mitotic arrest in the absence of spindle damage.
Cancer cell (2010-10-19)
Xing Zeng, Frederic Sigoillot, Shantanu Gaur, Sungwoon Choi, Kathleen L Pfaff, Dong-Chan Oh, Nathaniel Hathaway, Nevena Dimova, Gregory D Cuny, Randall W King
ABSTRACT
Microtubule inhibitors are important cancer drugs that induce mitotic arrest by activating the spindle assembly checkpoint (SAC), which, in turn, inhibits the ubiquitin ligase activity of the anaphase-promoting complex (APC). Here, we report a small molecule, tosyl-L-arginine methyl ester (TAME), which binds to the APC and prevents its activation by Cdc20 and Cdh1. A prodrug of TAME arrests cells in metaphase without perturbing the spindle, but nonetheless the arrest is dependent on the SAC. Metaphase arrest induced by a proteasome inhibitor is also SAC dependent, suggesting that APC-dependent proteolysis is required to inactivate the SAC. We propose that mutual antagonism between the APC and the SAC yields a positive feedback loop that amplifies the ability of TAME to induce mitotic arrest.
Product Description
proTAME, ≥90% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Nα-p-Tosyl-L-arginine methyl ester hydrochloride
Pricing and availability is not currently available.