Merck
  • Synthesis of affinity ligands and radioactive probes for isolation and study of myo-inositol 1,4,5-trisphosphate binding proteins.
Synthesis of affinity ligands and radioactive probes for isolation and study of myo-inositol 1,4,5-trisphosphate binding proteins.
Biochemistry (1990-05-29)
A N Jina, J Ralph, C E Ballou
ABSTRACT
To synthesize an affinity matrix for isolation of D-myo-inositol 1,4,5-trisphosphate binding proteins, racemic 3-cyclohexene-1-carboxaldehyde was oxidized and converted to a mixture of trans-3,4-di-hydroxycyclohexane-1-carboxylic acid methyl ester isomers, which was phosphorylated and separated into (+-)-(1R,3R,4R)- and (+-)-(1R,3S,4S)-trans-3,4-bis[(diphenoxyphosphoryl)oxy]cyclohex an e-1- carboxylic acid methyl esters. Each of these racemic compounds was hydrogenolyzed and reacted with ethylenediamine to give a monoamide, N-(2-aminoethyl)-bis(phosphonyloxy)cyclohexane-1-carboxamide, that was coupled to cyanogen bromide activated Sepharose 4B to provide the desired affinity matrices. The intermediate trans-3,4-bis[(diphenoxyphosphoryl)oxy]cyclohexane-1-carboxylic acid methyl ester was also reduced with lithium borotritide to give the (hydroxy[3H]methyl)cyclohexane derivative, which was phosphorylated and hydrogenolyzed to yield trans-3,4-bis(phosphonyloxy)-1-[(phosphonyloxy)[3H]methyl]cy clohexane, a radiolabeled analogue of inositol 1,4,5-trisphosphate. The carboxamide was also coupled to 4-azidosalicylic acid, and the product was iodinated to provide a 125I-radiolabeled photoactivatable cross-linking derivative of cyclohexanediol bisphosphate.
Sigma-Aldrich
Epoxy-activated−Sepharose 6B, lyophilized powder
Pricing and availability is not currently available.