Merck
  • The role of C-terminal part of ghrelin in pharmacokinetic profile and biological activity in rats.
The role of C-terminal part of ghrelin in pharmacokinetic profile and biological activity in rats.
Peptides (2011-02-05)
Naomi Morozumi, Takeshi Hanada, Hiromi Habara, Akira Yamaki, Mayumi Furuya, Takashi Nakatsuka, Norio Inomata, Yoshiharu Minamitake, Kazuhiro Ohsuye, Kenji Kangawa
ABSTRACT
Ghrelin is an endogenous ligand for growth hormone secretagogue receptor 1a (GHS-R1a), and consists of 28 amino acid residues with octanoyl modification at Ser(3). The previous studies have revealed that N-terminal part of ghrelin including modified Ser(3) is the active core for the activation of GHS-R1a. On the other hand, the role of C-terminal (8-28) region in ghrelin has not been clarified yet. In the present study, we prepared human ghrelin, C-terminal truncated ghrelin derivatives and anamorelin, a small molecular GHS compound which supposedly mimics the N-terminal active core, and examined GHS-R1a agonist activity in vitro, pharmacokinetic (PK) profile and growth hormone (GH) releasing activity in rats. All compounds demonstrated potent GHS-R1a agonist activities in vitro. Although the lack of C-terminal two amino acids did not modify PK profile and GH releasing activity, the deletion of C-terminal 8 and 20 amino acids affected them, and ghrelin(1-7)-Lys-NH(2) exhibited very short plasma half-life and low GH releasing activity in vivo. In rat plasma, ghrelin(1-7)-Lys-NH(2) was degraded more rapidly than ghrelin, suggesting that C-terminal part of ghrelin protected octanoylation of Ser(3) from plasma esterases. Subdiaphragmatic vagotomy significantly attenuated GH response to ghrelin but not to anamorelin. These results suggest that the C-terminal part of ghrelin has an important role in the biological activity in vivo. We also found that ghrelin stimulated GH release mainly via a vagal nerve pathway but anamorelin augmented GH release possibly by directly acting on brain in rats.
Product Description
Millipore
Rat/Mouse Growth Hormone ELISA, This Rat/Mouse Growth Hormone ELISA is used to measure & quantify Growth Hormone levels in Metabolism & Endocrine & Cell Signaling research.
Pricing and availability is not currently available.