Merck
  • Role of the beta-subunit arginine/lysine finger in integrin heterodimer formation and function.
Role of the beta-subunit arginine/lysine finger in integrin heterodimer formation and function.
Journal of immunology (Baltimore, Md. : 1950) (2008-01-23)
Vineet Gupta, José Luis Alonso, Takashi Sugimori, Makram Essafi, Makram Issafi, Jiang-Ping Xiong, M Amin Arnaout
ABSTRACT
Formation of the integrin alphabeta heterodimer is essential for cell surface expression and function. At the core of the alphabeta interface is a conserved Arg/Lys "finger" from the beta-subunit that inserts into a cup-like "cage" formed of two layers of aromatic residues in the alpha-subunit. We evaluated the role of this residue in heterodimer formation in an alphaA-lacking and an alphaA-containing integrin alphaVbeta3 and alphaMbeta2 (CD11b/CD18), respectively. Arg261 of beta3 was mutated to Ala or Glu; the corresponding Lys252 of beta2 was mutated to Ala, Arg, Glu, Asp, or Phe; and the effects on heterodimer formation in each integrin examined by ELISA and immunoprecipitation in HEK 293 cells cotransfected with plasmids encoding the alpha- and beta-subunits. The Arg261Glu (but not Arg261Ala) substitution significantly impaired cell surface expression and heterodimer formation of alphaVbeta3. Although Lys252Arg, and to a lesser extent Lys252Ala, were well tolerated, each of the remaining substitutions markedly reduced cell surface expression and heterodimer formation of CD11b/CD18. Lys252Arg and Lys252Ala integrin heterodimers displayed a significant increase in binding to the physiologic ligand iC3b. These data demonstrate an important role of the Arg/Lys finger in formation of a stable integrin heterodimer, and suggest that subtle changes at this residue affect the activation state of the integrin.
Sigma-Aldrich
Anti-Integrin β3 Antibody, clone PM6/13, clone PM6/13, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin alphaV Antibody, CT, Intracellular, serum, Chemicon®