Merck
  • Tumor-suppressive microRNA-145 induces growth arrest by targeting SENP1 in human prostate cancer cells.
Tumor-suppressive microRNA-145 induces growth arrest by targeting SENP1 in human prostate cancer cells.
Cancer science (2015-02-04)
Chunyang Wang, Weiyang Tao, Shaobin Ni, Qiyin Chen, Zhongshan Zhao, Li Ma, Yiming Fu, Zhixing Jiao
ABSTRACT
Prostate cancer (PCa) prevails as the most commonly diagnosed malignancy in men and the third leading cause of cancer-related deaths in developed countries. One of the distinct characteristics of prostate cancer is overexpression of the small ubiquitin-like modifier (SUMO)-specific protease 1 (SENP1), and the upregulation of SENP1 contributes to the malignant progression and cell proliferation of PCa. Previous studies have shown that the expression of microRNA-145 (miRNA-145) was extensively deregulated in PCa cell lines and primary clinical prostate cancer samples. Independent target prediction methods have indicated that the 3'-untranslated region of SENP1 mRNA is a potential target of miR-145. Here we found that low expression of miR-145 was correlated with high expression of SENP1 in PCa cell line PC-3. The transient introduction of miR-145 caused cell cycle arrest in PC-3 cells, and the opposite effect was observed when miR-145 inhibitor was transfected. Further studies revealed that the SENP1 3'-untranslated region was a regulative target of miR-145 in vitro. MicroRNA-145 also suppressed tumor formation in vivo in nude mice. Taken together, miR-145 plays an important role in tumorigenesis of PCa through interfering SENP1.
Sigma-Aldrich
Anti-Ki-67 Antibody, Chemicon®, from rabbit
Pricing and availability is not currently available.
Sigma-Aldrich
Rabbit anti-GAPDH Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
Pricing and availability is not currently available.