Merck
  • Identification and role of muscarinic receptor subtypes expressed in rat adrenal medullary cells.
Identification and role of muscarinic receptor subtypes expressed in rat adrenal medullary cells.
Journal of pharmacological sciences (2011-11-19)
Keita Harada, Hidetada Matsuoka, Takeyoshi Sata, Akira Warashina, Masumi Inoue
ABSTRACT
The muscarinic receptor is known to be involved in the acetylcholine (ACh)-induced secretion of catecholamines in the adrenal medullary (AM) cells of various mammals. The muscarinic receptor subtype involved and its physiological role, however, have not been elucidated yet. Thus, we investigated these issues in acutely isolated rat AM cells and perfused rat adrenal medulla. The RT-PCR analysis revealed the presence of M(2), M(3), M(4), and M(5) mRNAs. Immunocytochemistry with specific antibodies showed that M(5)-like immunoreactivities (IRs) were detected at half the cell membrane area, which was much larger than that with M(3)- or M(4)-like IRs. Muscarine produced inward currents in a dose-dependent manner. Pilocarpine, McN-A-343, and oxotremorine were less efficient than muscarine; and RS-86, which has no action on the M(5) receptor, produced no current. Electrical stimulation of nerve fibers produced a frequency-dependent increase in the Ca(2+) signal in perfused adrenal medullae. Muscarinic receptors were found to be involved in neuronal transmission in AM cells in the presence of a cholinesterase inhibitor, which suppresses ACh degradation. We concluded that the M(5) receptor is the major muscarinic receptor subtype in rat AM cells and may be involved in neuronal transmission under conditions where ACh spills over the synapse.
Sigma-Aldrich
Anti-Actin Antibody, clone C4, clone C4, Chemicon®, from mouse
Sigma-Aldrich
Anti-CHAPSYN-110 Antibody, Chemicon®, from rabbit