- Mechanisms of enzymatic glycoside hydrolysis.
Mechanisms of enzymatic glycoside hydrolysis.
Current opinion in structural biology (1994-12-01)
J D McCarter, S G Withers
PMID7712292
ABSTRACT
The determination of a large number of three-dimensional structures of glycosidases, both free and in complex with ligands, has provided valuable new insights into glycosidase catalysis, especially when coupled with results from studies of specifically labelled glycosidases and kinetic analyses of point mutants.
Sigma-Aldrich
β-Glucuronidase from limpets (Patella vulgata), Type L-II, lyophilized powder, 1,000,000-3,000,000 units/g solid