Merck
  • Substances with alpha-melanotropin-like immunoreactivity in bovine brains.
Substances with alpha-melanotropin-like immunoreactivity in bovine brains.
The International journal of biochemistry (1992-04-01)
T B Ng
ABSTRACT
1. Bovine cerebral hemispheres were extracted with an acidic medium (acetone-water-hydrochloric acid mixture, 40:5:1 by volume, pH 1.8). The precipitate which formed upon addition of a copious volume of cold acetone to the extract was designated acid acetone powder (AAP). 2. The AAP was then subjected to ion exchange chromatography on carboxymethyl (CM)-cellulose, gel filtration on Sephadex G100 and Sephadex G25, second ion exchange chromatography on CM-cellulose and high performance liquid chromatography. The absorbance of all fractions was measured at 280 nm and their alpha-melanotropin-(alpha-MSH)-like immunoreactivity was monitored with radioimmunoassay. 3. It was found that alpha-MSH-like immunoreactivity and bioactivity (lipolytic activity) was due to low molecular weight materials as evidenced by their retardation on Sephadex G-100 and Sephadex G-25. The immunoreactivity was distributed among fractions adsorbed and fractions unadsorbed on CM-cellulose and also among high performance liquid chromatographic fractions signifying the presence of multiple alpha-MSH-like molecules.