Merck
  • Optical tissue clearing in combination with perfusion and immunofluorescence for placental vascular imaging.
Optical tissue clearing in combination with perfusion and immunofluorescence for placental vascular imaging.
Medicine (2018-10-04)
Maira Carrillo, Marcel Chuecos, Kushal Gandhi, Andrey Bednov, David Lee Moore, James Maher, Gary Ventolini, Guangchen Ji, Natalia Schlabritz-Loutsevitch
ABSTRACT
Imaging of placental tissues is a difficult task, because of specific for this organ complex multicellular and 3D tissue structure. The tissue clearing systems (X-CLARITY) system is a valuable tool for the examining the expression of molecular pathways in whole tissues and organs, originally developed for brain imaging.In the present report, we utilized this technology for the examination of placental vasculature and protein expression in perfused human placental tissue.The placental tissue was sufficiently cleared with preservation of endothelial staining and fluorescent markers, allowing visualization using confocal microscopy. The CLARITY method and X-CLARITY system is a valuable tool in placental imaging.
Sigma-Aldrich
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine perchlorate, BioReagent, suitable for fluorescence, ≥98.0% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 60,000-76,000
Pricing and availability is not currently available.