Merck
  • [omim][BF4]-mediated toxicity in mussel hemocytes includes its interaction with cellular membrane proteins.
[omim][BF4]-mediated toxicity in mussel hemocytes includes its interaction with cellular membrane proteins.
Aquatic toxicology (Amsterdam, Netherlands) (2018-08-14)
Vasiliki Tsarpali, Stefanos Dailianis
ABSTRACT
The current study is based on the increasing demand for the assessment of ionic liquid (IL)-mediated aquatic toxicity. Specifically, although a lot of studies have been performed so far, investigating IL-mediated adverse effects on numerous aquatic organisms, little is known about their mode of action. Given that the use of in vitro models is considered as a reliable tool for determining the mediated biological effects, the modulation of specific biochemical pathways and the onset of various forms of damage with great precision and reproducibility, mixed primary cultures of mussel Mytilus galloprovincialis hemocytes were used for investigating whether 1-octyl-3-methylimidazolium tetrafluoroborate ([omim][BF4]) mediated toxicity is related to its interaction with cellular membrane proteins. Specifically, [omim][BF4]-mediated cytotoxic, oxidative and genotoxic effects were investigated in mussel hemocytes before and after pre-treatment of cells with non-toxic concentration of guanidine hydrochloride (1 mM GndHCl). The results showed that [omim][BF4] at concentrations ranging from 0.7 to 1.75 μM can induce cytotoxic (almost <50% reduction of cell viability), oxidative (increased levels of O2•- production and lipid peroxidation by-products) and genotoxic (increased levels of DNA damage) effects, while cells pre-treated with 1 mM GndHCl showed a significant attenuation of IL's toxic potency in all cases. According to the latter, the current study showed that [omim][BF4]-mediated toxicity could be related not only to its well-known interaction with membrane lipid bilayers, but also to its interference with membrane proteins. Using GndHCl, a chaotropic agent that disrupts the hydrogen bonding network and the stability of membrane proteins via its interference with the intramolecular interactions mediated by non-covalent forces on cellular membranes, it was firstly shown that altering the membrane integrity as well as the native state of cellular membrane proteins, by weakening the hydrophobic effect, could attenuate the possible interaction of [omim][BF4] with cellular membranes and the concomitant induction of protein-based intracellular processes, commonly linked with the induction of severe cellular damage.
Sigma-Aldrich
1-Methyl-3-octylimidazolium tetrafluoroborate, ≥97.0% (HPLC)
Pricing and availability is not currently available.