Merck
  • 27-Hydroxycholesterol and 7alpha-hydroxycholesterol trigger a sequence of events leading to migration of CCR5-expressing Th1 lymphocytes.
27-Hydroxycholesterol and 7alpha-hydroxycholesterol trigger a sequence of events leading to migration of CCR5-expressing Th1 lymphocytes.
Toxicology and applied pharmacology (2013-12-29)
Sun-Mi Kim, Bo-Young Kim, Sae-A Lee, Seong-Kug Eo, Yungdae Yun, Chi-Dae Kim, Koanhoi Kim
ABSTRACT
Th1 lymphocytes are predominant in atherosclerotic lesions. However, mechanisms involved in the Th1 predominance are unknown. We have investigated the possibility of Th1 lymphocyte recruitment in a cholesterol-rich milieu. A high cholesterol diet resulted in enhanced expression of CCR5 ligands, including CCL3 and CCL4, but not of proatherogenic CXCR3 ligands, in atherosclerotic arteries of ApoE(-/-) mice. 27-Hydroxycholesterol and 7α-hydroxycholesterol, cholesterol oxides (oxysterols) detected in abundance in atherosclerotic lesions, greatly induced the transcription of CCL3 and CCL4 genes in addition to enhancing secretion of corresponding proteins by THP-1 monocytic cells. However, an identical or even higher concentration of cholesterol, 7β-hydroxycholesterol, and 7-ketocholsterol did not influence expression of these chemokines. Conditioned media containing the CCR5 ligands secreted from THP-1 cells induced migration of Jurkat T cells expressing CCR5, a characteristic chemokine receptor of Th1 cells, but not of Jurkat T cells that do not express CCR5. The migration of CCR5-expressing Jurkat T cells was abrogated in the presence of a CCR5-neutralizing antibody. 27-Hydroxycholesterol and 7α-hydroxycholesterol enhanced phosphorylation of Akt. Pharmacological inhibitors of phosphoinositide-3-kinase/Akt pathways blocked transcription as well as secretion of CCL3 and CCL4 in conjunction with attenuated migration of CCR5-expressing Jurkat T cells. This is the first report on the involvement of cholesterol oxides in migration of distinct subtype of T cells. We propose that 27-hydroxycholesterol and 7α-hydroxycholesterol can trigger a sequence of events that leads to recruitment of Th1 lymphocytes and phosphoinositide-3-kinase/Akt pathways play a major role in the process.
Product Description
Avanti
27-hydroxycholesterol, Avanti Research - A Croda Brand
Pricing and availability is not currently available.