Merck
Ammonia as a pheromone in anuran tadpoles.
Physiological research (2011-07-23)
Y B Manteifel, E I Kiseleva
ABSTRACT
In laboratory experiments, conspecific excretions and ammonia solutions evoked avoidance reactions in tadpoles of three anuran species, Bufo bufo, Rana temporaria, and Rana arvalis. A differential sensitivity of ammonia chemoreception was determined for two anuran species. For Bufo bufo tadpoles, these characteristics at ammonia background concentration of 0.2 mg/l lied in the range 150 % < dI/I < 500 %, and for background of 0.4 mg/l the value lied in the range 400 % < dI/I < 500 %. For Rana temporaria tadpoles, differential threshold against ammonia background concentration of 0.15 mg/l was close to 200 % and against background ammonia concentration of 1.1 mg/l was close to 100 %. These results suggest that such sensitivity of both anurans is sufficient for using ammonia in intra- and interspecies communication.
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Pricing and availability is not currently available.