Merck
  • Quercetin‑3‑O‑β‑D‑glucoside decreases the bioavailability of cyclosporin A through regulation of drug metabolizing enzymes, transporters and nuclear receptors in rats.
Quercetin‑3‑O‑β‑D‑glucoside decreases the bioavailability of cyclosporin A through regulation of drug metabolizing enzymes, transporters and nuclear receptors in rats.
Molecular medicine reports (2018-07-18)
Tingyu Yang, Yani Liu, Xixi Huang, Rui Zhang, Chunxiao Yang, Jiali Zhou, Yu Zhang, Jing Wan, Shaojun Shi
ABSTRACT
Quercetin is a flavonoid compound that is widely present in food and drink. Quercetin‑3‑O‑β‑D‑glucoside (Q3GA) is a major metabolite of quercetin. The aim of the present study was to investigate the effect of Q3GA on the pharmacokinetics of orally and intravenously administered cyclosporin A (CsA) in rats, and to assess the effect of Q3GA on drug‑metabolizing enzymes (DMEs), drug transporters (DTs) and nuclear receptors (NRs). The pharmacokinetic parameters of CsA were measured following oral (10 mg/kg) and intravenous (2.5 mg/kg) administration of CsA in the presence or absence of Q3GA. The mRNA and protein expression levels of DMEs, DTs and NRs in the liver and small intestine were detected by quantitative polymerase chain reaction and western blot analysis. The results indicated that the intravenous administration of Q3GA (2.5, 5 or 10 mg/kg) for 7 consecutive days reduced the bioavailability of oral CsA. By contrast, the pharmacokinetics of the intravenous administration of CsA were not affected by Q3GA. However, the mRNA and protein expression levels of DMEs and DTs were inhibited by Q3GA. The activation of DMEs and DTs by NRs, and the interplay between DMEs and DTs, may explain these results. The present study identified a novel flavonoid‑drug interaction, which may have implications for patients taking CsA and quercetin supplements or on a quercetin‑containing diet.
Sigma-Aldrich
3-Hydroxyflavone, ≥98%