Merck
  • Azilsartan improves the effects of etanercept in patients with active rheumatoid arthritis: a pilot study.
Azilsartan improves the effects of etanercept in patients with active rheumatoid arthritis: a pilot study.
Therapeutics and clinical risk management (2018-08-21)
Naza Mohammed Ali Mahmood, Saad Abdulrahman Hussain, Raouf Rahim Mirza
ABSTRACT
Much evidence has emerged documenting the involvement of the renin-angiotensin system (RAS) in inflammatory processes. The objective of this study was to evaluate the effects of blocking RAS with azilsartan (Azil) on the clinical efficacy of etanercept (Etan) in patients with active rheumatoid arthritis (RA). Forty-two patients diagnosed with active RA and poorly responding to methotrexate were enrolled in this pilot clinical study. They were randomly allocated into two groups, and treated with either Etan (50 mg/week) and placebo or the same dose of Etan with Azil (20 mg/day) for 90 days. The clinical outcome was evaluated using the Disease Activity Score-28 joint (DAS-28), simplified disease activity index (SDAI), clinical disease activity index (CDAI) and the health assessment questionnaire disease index (HAQ-DI). Blood samples were obtained for the assessment of C-reactive protein and erythrocyte sedimentation rate at baseline and after 90 days. The markers of pain and disease activity, C-reactive protein and erythrocyte sedimentation rate were significantly improved when Azil was used, as an adjuvant with Etan, compared with the use of Etan and placebo. Blocking RAS with azilsartan may improve the effects of etanercept on the clinical markers of pain and disease severity of patients with active RA not responding to methotrexate.
Sigma-Aldrich
Azilsartan, ≥98% (HPLC)