- Acyclic diaminocarbenes: simple, versatile ligands for cross-coupling reactions.
Acyclic diaminocarbenes: simple, versatile ligands for cross-coupling reactions.
Chemical communications (Cambridge, England) (2006-02-01)
Bhartesh Dhudshia, Avinash N Thadani
PMID16446846
ABSTRACT
Acyclic diaminocarbenes are found to be useful ligands for palladium catalyzed Suzuki-Miyaura, Sonogashira and Heck cross-coupling reactions of aryl/alkenyl bromides and chlorides.