Merck
Interrogating the ccm-3 Gene Network.
Cell reports (2018-09-13)
Benjamin Lant, Swati Pal, Eric Michael Chapman, Bin Yu, Daniel Witvliet, Soo Choi, Lisa Zhao, Corinne Albiges-Rizo, Eva Faurobert, W Brent Derry
ABSTRACT
Cerebral cavernous malformations (CCMs) are neurovascular lesions caused by mutations in one of three genes (CCM1-3). Loss of CCM3 causes the poorest prognosis, and little is known about how it regulates vascular integrity. The C. elegans ccm-3 gene regulates the development of biological tubes that resemble mammalian vasculature, and in a genome-wide reverse genetic screen, we identified more than 500 possible CCM-3 pathway genes. With a phenolog-like approach, we generated a human CCM signaling network and identified 29 genes in common, of which 14 are required for excretory canal extension and membrane integrity, similar to ccm-3. Notably, depletion of the MO25 ortholog mop-25.2 causes severe defects in tube integrity by preventing CCM-3 localization to apical membranes. Furthermore, loss of MO25 phenocopies CCM3 ablation by causing stress fiber formation in endothelial cells. This work deepens our understanding of how CCM3 regulates vascular integrity and may help identify therapeutic targets for treating CCM3 patients.
Sigma-Aldrich
Trypsin inhibitor from Glycine max (soybean), powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Pricing and availability is not currently available.