Merck
  • Cartilage oligomeric matrix protein is a novel notch ligand driving embryonic stem cell differentiation towards the smooth muscle lineage.
Cartilage oligomeric matrix protein is a novel notch ligand driving embryonic stem cell differentiation towards the smooth muscle lineage.
Journal of molecular and cellular cardiology (2018-07-08)
Baihui Ma, Fang Yao, Nan Xie, Chenfeng Mao, Fei Liu, Ze Gong, Guizhen Zhao, Zhujiang Liu, Zeyu Cai, Fang Yu, Rongbo Dai, Zhongjiang Chen, Li Wang, Qingbo Xu, Wei Kong, Yi Fu
ABSTRACT
Cartilage oligomeric matrix protein (COMP), a protective component of vascular extracellular matrix (ECM), maintains the homeostasis of mature vascular smooth muscle cells (VSMCs). However, whether COMP modulates the differentiation of stem cells towards the smooth muscle lineage is still elusive. Firstly, purified mouse COMP directly induced mouse embryonic stem cell (ESC) differentiation into VSMCs both in vitro and in vivo, while the silencing of endogenous COMP markedly inhibited ESC-VSMC differentiation. RNA-Sequencing revealed that Notch signaling was significantly activated by COMP during ESC-VSMC differentiation, whereas the inhibition of Notch signaling attenuated COMP-directed ESC-VSMC differentiation. Furthermore, COMP deficiency inhibited Notch activation and VSMC differentiation in mice. Through silencing distinct Notch receptors, we identified that Notch1 mainly mediated COMP-initiated ESC-VSMC differentiation. Mechanistically, COMP N-terminus directly interacted with the EGF11-12 domain of Notch1 and activated Notch1 signaling, as evidenced by co-immunoprecipitation and mammalian two-hybrid assay. In conclusion, COMP served as a potential ligand of Notch1, thereby driving ESC-VSMC differentiation.
Sigma-Aldrich
Heparin−Agarose, (1:1 suspension in a 20% ethanol solution)
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid