Merck
  • Phospholipase D1 Acts through Akt/TopBP1 and RB1 to Regulate the E2F1-Dependent Apoptotic Program in Cancer Cells.
Phospholipase D1 Acts through Akt/TopBP1 and RB1 to Regulate the E2F1-Dependent Apoptotic Program in Cancer Cells.
Cancer research (2016-10-30)
Dong Woo Kang, Shin Wha Lee, Won Chan Hwang, Bo Hui Lee, Yong-Seok Choi, Young-Ah Suh, Kang-Yell Choi, Do Sik Min
ABSTRACT
The RB1/E2F1 signaling pathway is frequently deregulated in colorectal cancer and has been suggested to intersect with Wnt/β-catenin and PI3K/Akt pathways, but molecular evidence for this link is lacking. In this study, we demonstrate that phospholipase D1 (PLD1), a transcriptional target of β-catenin/TCF4, orchestrates functional interactions between these pathways during intestinal tumor development. Overexpression of PLD1 in intestinal epithelial cells protected cells from apoptosis induced by PLD1 ablation in the Apcmin/+ mouse model of intestinal tumorigenesis. Mechanistic investigations revealed that genetic and pharmacologic targeting of PLD1 promote the E2F1-dependent apoptotic program via both miR-192/4465-mediated downregulation of RB1 and inhibition of Akt-TopBP1 pathways. Moreover, the miRNA-RB1 axis and Akt pathway also contributed to the PLD1-mediated self-renewal capacity of colon cancer-initiating cells. Finally, PLD1-driven E2F1 target gene expression positively correlated with tumor stage in patients with colorectal cancer. Overall, our findings suggest that PLD1 mediates cross-talk between multiple major signaling pathways to promote the survival and malignancy of colon cancer cells and may therefore represent an ideal signaling node for therapeutic targeting. Cancer Res; 77(1); 142-52. ©2016 AACR.
Sigma-Aldrich
VU0155069, ≥98% (HPLC)
Pricing and availability is not currently available.