Merck
  • Modulating D-amino acid oxidase (DAAO) substrate specificity through facilitated solvent access.
Modulating D-amino acid oxidase (DAAO) substrate specificity through facilitated solvent access.
PloS one (2018-06-16)
Kalyanasundaram Subramanian, Artur Góra, Ruud Spruijt, Karolina Mitusińska, Maria Suarez-Diez, Vitor Martins Dos Santos, Peter J Schaap
ABSTRACT
D-amino acid oxidase (DAAO) degrades D-amino acids to produce α-ketoacids, hydrogen peroxide and ammonia. DAAO has often been investigated and engineered for industrial and clinical applications. We combined information from literature with a detailed analysis of the structure to engineer mammalian DAAOs. The structural analysis was complemented with molecular dynamics simulations to characterize solvent accessibility and product release mechanisms. We identified non-obvious residues located on the loops on the border between the active site and the secondary binding pocket essential for pig and human DAAO substrate specificity and activity. We engineered DAAOs by mutating such critical residues and characterised the biochemical activity of the resulting variants. The results highlight the importance of the selected residues in modulating substrate specificity, product egress and enzyme activity, suggesting further steps of DAAO re-engineering towards desired clinical and industrial applications.
Sigma-Aldrich
D-Penicillamine, 98-101%
Pricing and availability is not currently available.