Merck
  • Biological screening of extracts from leaf and stem bark of Croton floribundus Spreng. (Euphorbiaceae).
Biological screening of extracts from leaf and stem bark of Croton floribundus Spreng. (Euphorbiaceae).
Brazilian journal of biology = Revista brasleira de biologia (2018-01-11)
E F Barth, L S Pinto, P Dileli, D C Biavatti, Y L Silva, W Bortolucci, Z C Gazim, O S Takemura, M B Romagnolo, A Laverde-Junior
ABSTRACT
This work describes the preliminary evaluation of cytotoxic, antimicrobial, molluscicidal, antioxidant and anticholinesterase activities from leaf (LECF) and stem bark alcoholic extracts (BECF) of the species Croton floribundus Spreng. (Euphorbiaceae), popularly known as capixingui or tapixingui. BECF presented significant toxicity (LC50 = 89.6 μg/ml) in the Artemia salina Leach, 1819 (Crustacea: Branchiopoda) bioassay, whereas LECF did not show activity (LC50 > 1000 μg/ml). From DPPH method, the values of IC50 for the LECF and BECF were 61.2 μg/ml and 62.2 μg/ml, respectively, showing that C. floribundus has an expressive antioxidant activity. Antimicrobial susceptibility was evaluated by microdilution technique and only BECF was active against Staphylococcus aureus (MIC = 39.6 μg/ml). The extracts did not present molluscicidal activity against snail Biomphalaria glabrata Say, 1818 (Gastropoda: Planorbidae). Both extracts revealed the presence of several components with an inhibiting capacity of acetylcholinesterase enzyme on the bioautographic assay. C. floribundus showed to be a promising species considering that it exhibited good biological activity in the most assays performed.
Sigma-Aldrich
1-Naphthyl acetate, ≥98% (C)