Merck
  • UVΑ pre-irradiation to P25 titanium dioxide nanoparticles enhanced its toxicity towards freshwater algae Scenedesmus obliquus.
UVΑ pre-irradiation to P25 titanium dioxide nanoparticles enhanced its toxicity towards freshwater algae Scenedesmus obliquus.
Environmental science and pollution research international (2018-04-04)
Barsha Roy, Hemamalini Chandrasekaran, Suresh Palamadai Krishnan, Natarajan Chandrasekaran, Amitava Mukherjee
ABSTRACT
There has recently been an increase in the usage of TiO2 nanoparticles (NPs). P25 TiO2 NPs, a mixture of anatase and rutile phase in 3:1 ratio, are generally used for photocatalytic applications because both phases exhibit a synergistic effect on the photocatalytic activity of the TiO2 NPs. In the present study, increased toxicity of UVA-pre-irradiated P25 TiO2 NPs on freshwater algae Scenedesmus obliquus was assessed under visible light and dark exposure conditions at actual low concentrations (0.3, 3 and 35 μM of Ti). Photocatalytic property of P25 TiO2 NPs caused disaggregation of UVA-pre-irradiated NPs, thus significantly decreasing the mean hydrodynamic diameter (MHD) (188.74 ± 0.54 nm) than that of non-irradiated NPs (232.26 ± 0.44). This decrease in diameter of UVA-pre-irradiated NPs may increase its biological activity towards algal samples. All concentrations of pre-irradiated NPs, under both light and dark conditions, showed a significantly lesser cell viability (p < 0.001) when compared with non-irradiated NPs. Increased production of ROS, antioxidant enzymes and lipid peroxidation supported the viability data. Higher exopolysaccharide production and more nuclear damage were observed for pre-irradiated NPs. NP uptake was also more for the pre-irradiated NPs on treated samples when compared with non-irradiated NPs on treated samples, which, in turn, established the higher toxic potential of UVA-pre-irradiated TiO2 NPs. This study improves our understanding of the toxic effects of UVA-pre-irradiated TiO2 NPs on freshwater algae, thereby emphasising the need for ecological risk assessments of metal oxide nanoparticles in a natural experimental medium.
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)