Merck
  • Functionalized carbon dot adorned coconut shell char derived green catalysts for the rapid synthesis of amidoalkyl naphthols.
Functionalized carbon dot adorned coconut shell char derived green catalysts for the rapid synthesis of amidoalkyl naphthols.
Journal of colloid and interface science (2018-03-13)
Divya P Narayanan, Sudha Kochiyil Cherikallinmel, Sugunan Sankaran, Binitha N Narayanan
ABSTRACT
A one pot synthesis of carbon dot incorporated porous coconut shell char derived sulphonated catalyst is reported here for the first time and is effectively used in the multicomponent synthesis of amidoalkyl naphthol. Macroporous nature of the char is revealed from scanning electron microscopic (SEM) analysis, whereas the dispersion of carbon dots (CDs) on the porous coconut shell char is confirmed from the high resolution transmission electron microscopic (HRTEM) analysis. Fluorescence emission spectrum further confirmed the presence of CDs in the catalyst. Fourier-transform infrared (FTIR) spectral analysis of the materials indicated that sulphonation occurred both to the CD and to the porous char. X-ray photo electron spectroscopic (XPS) analysis of the most active catalyst confirmed the presence of both sulphonic acid and carboxylic acid groups in the catalyst. The coconut shell char derived materials prepared by varying the amount of H2SO4 are successfully utilized as efficient alternative green catalysts for the multicomponent reaction, where excellent activity in amidoalkyl naphthol synthesis is obtained within short periods under solvent free reaction conditions. A maximum yield of 98% is obtained in the synthesis of N-[Phenyl-(2-hydroxy-naphthalen-1-yl)-methyl]-benzamide, the representative amidoalkyl naphthol, with the best catalyst within 3 min of reaction. The catalyst is highly active for the reactions carried out with varieties of aldehydes and amides with a product yield in the range of 88-98%. The best catalyst system retained more than 90% of its initial activity even upto 6th repeated run.
Sigma-Aldrich
4-Chlorobenzaldehyde, 97%
Pricing and availability is not currently available.