Merck
  • Efficient (3S)-Acetoin and (2S,3S)-2,3-Butanediol Production from meso-2,3-Butanediol Using Whole-Cell Biocatalysis.
Efficient (3S)-Acetoin and (2S,3S)-2,3-Butanediol Production from meso-2,3-Butanediol Using Whole-Cell Biocatalysis.
Molecules (Basel, Switzerland) (2018-03-23)
Yuanzhi He, Feixue Chen, Meijing Sun, Huifang Gao, Zewang Guo, Hui Lin, Jiebo Chen, Wensong Jin, Yunlong Yang, Liaoyuan Zhang, Jun Yuan
ABSTRACT
(3S)-Acetoin and (2S,3S)-2,3-butanediol are important platform chemicals widely applied in the asymmetric synthesis of valuable chiral chemicals. However, their production by fermentative methods is difficult to perform. This study aimed to develop a whole-cell biocatalysis strategy for the production of (3S)-acetoin and (2S,3S)-2,3-butanediol from meso-2,3-butanediol. First, E. coli co-expressing (2R,3R)-2,3-butanediol dehydrogenase, NADH oxidase and Vitreoscilla hemoglobin was developed for (3S)-acetoin production from meso-2,3-butanediol. Maximum (3S)-acetoin concentration of 72.38 g/L with the stereoisomeric purity of 94.65% was achieved at 24 h under optimal conditions. Subsequently, we developed another biocatalyst co-expressing (2S,3S)-2,3-butanediol dehydrogenase and formate dehydrogenase for (2S,3S)-2,3-butanediol production from (3S)-acetoin. Synchronous catalysis together with two biocatalysts afforded 38.41 g/L of (2S,3S)-butanediol with stereoisomeric purity of 98.03% from 40 g/L meso-2,3-butanediol. These results exhibited the potential for (3S)-acetoin and (2S,3S)-butanediol production from meso-2,3-butanediol as a substrate via whole-cell biocatalysis.
Sigma-Aldrich
(2R,3R)-(−)-2,3-Butanediol, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
meso-2,3-Butanediol, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
N-Octanoyl-N-methylglucamine, ≥97% (GC)
Pricing and availability is not currently available.