Merck
  • Toxicity of Bifenthrin and Mixtures of Bifenthrin Plus Acephate, Imidacloprid, Thiamethoxam, or Dicrotophos to Adults of Tarnished Plant Bug (Hemiptera: Miridae).
Toxicity of Bifenthrin and Mixtures of Bifenthrin Plus Acephate, Imidacloprid, Thiamethoxam, or Dicrotophos to Adults of Tarnished Plant Bug (Hemiptera: Miridae).
Journal of economic entomology (2018-01-24)
Moneen M Jones, Jessica L Duckworth, Jacqueline Robertson
ABSTRACT
To assess the toxicity of bifenthrin and four mixtures of insecticides to tarnished plant bug, we used an insecticide dip method of green bean to treat adults of a laboratory colony; mortality was assessed after 48 h. LC50s for imidacloprid, bifenthrin, acephate, thiamethoxam, and dicrotophos were 0.12, 0.39, 0.62, 0.67, and 3.96 ppm, respectively. LC75s for imidacloprid, bifenthrin, acephate, thiamethoxam, and dicrotophos were 0.61, 4.22, 5.10, 2.65, and 7.86 ppm, respectively. Based on the LC50s and LC75s, dicrotophos was much less toxic than the other chemicals tested. PoloMix software was used to determine syngerism, antagonism, or addition effects of the mixtures. Three out of four analyses of the joint action of bifenthrin plus imidacloprid or acephate or dicrotophos showed that toxicity was not independent and not correlated. For bifenthrin plus dicrotophos, observed mortality was greater than expected mortality at most concentrations suggesting synergism. Mixtures of bifenthrin plus imidacloprid and bifenthrin plus acephate showed observed mortality significantly less than expected, suggesting antagonism. LC50s for bifenthrin plus dicrotophos, acephate, imidacloprid, and thiamethoxam were 0.38, 1.06, 0.17, and 0.26 ppm, respectively. LC75s for bifenthrin plus dicrotophos, acephate, imidacloprid, and thiamethoxam were 13.61, 13.18, 0.67, and 0.80 ppm, respectively. Based on the LC50s and LC75s, bifenthrin plus acephate was 3- to 10-fold less toxic than the other chemicals tested. Bifenthrin plus acephate is frequently used in tank mixes to control tarnished plant bug and other cotton pests, and the effectiveness of each individual chemical appears to be reduced in one to one ratio mixtures.
Product Description
Supelco
Thiamethoxam, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Supelco
Dicrotophos, PESTANAL®, analytical standard
Pricing and availability is not currently available.