MilliporeSigma
  • Novel lactonization of ethenetricarboxylate derivatives: intermolecular trapping of alkenes.
Novel lactonization of ethenetricarboxylate derivatives: intermolecular trapping of alkenes.
Organic letters (2005-02-25)
Shoko Yamazaki, Kanae Ohmitsu, Kunihiro Ohi, Tetsuya Otsubo, Kayo Moriyama
ABSTRACT
A novel cyclization of 1,1-diethyl 2-tert-butyl ethenetricarboxylate (1a) in the presence of a Lewis acid afforded a 5,5-dimethyl-gamma-lactone derivative 2a. The reaction process has been shown to arise from formation by trapping of isobutylene generated in situ. Lewis acid-promoted intermolecular reactions of 1,1-diethyl 2-hydrogen ethenetricarboxylate (5) and various alkenes to afford highly functionalized gamma-lactones were also developed. [reaction: see text]