Merck
  • Development and characterisation of a low-concentration sodium dodecyl sulphate decellularised porcine dermis.
Development and characterisation of a low-concentration sodium dodecyl sulphate decellularised porcine dermis.
Journal of tissue engineering (2017-08-18)
Jack A Helliwell, Daniel S Thomas, Vaia Papathanasiou, Shervanthi Homer-Vanniasinkam, Amisha Desai, Louise M Jennings, Paul Rooney, John N Kearney, Eileen Ingham
ABSTRACT
The aim of this study was to adapt a proprietary decellularisation process for human dermis for use with porcine skin. Porcine skin was subject to: sodium chloride (1 M) to detach the epidermis, trypsin paste to remove hair follicles, peracetic acid (0.1% v/v) disinfection, washed in hypotonic buffer and 0.1% (w/v) sodium dodecyl sulphate in the presence of proteinase inhibitors followed by nuclease treatment. Cellular porcine skin, decellularised porcine and human dermis were compared using histology, immunohistochemistry, GSL-1 lectin (alpha-gal epitope) staining, biochemical assays, uniaxial tensile and in vitro cytotoxicity tests. There was no microscopic evidence of cells in decellularised porcine dermis. DNA content was reduced by 98.2% compared to cellular porcine skin. There were no significant differences in the biomechanical parameters studied or evidence of cytotoxicity. The decellularised porcine dermis retained residual alpha-gal epitope. Basement membrane collagen IV immunostaining was lost following decellularisation; however, laminin staining was retained.
Sigma-Aldrich
Monoclonal Anti-Laminin antibody produced in mouse, clone LAM-89, ascites fluid
Pricing and availability is not currently available.